Order Entry
Northern Ireland
ContactUsLinkComponent
Bathocuproin 98%
Irritant
Bathocuproin 98%
Certificates
undefined
Irritant
Bathocuproin 98%

Specification Test Results

  • Appearance:
    White to cream to yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    277 to 285 °C (Dec)
  • Titration with HClO4:
    97.5 to 102.5 %

Specification Test Results

Related Information

Frequently Bought Together