ContactUsLinkComponent
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride 99%
undefined
2-(1-Naphthylmethyl)-2-imidazoline hydrochloride 99%
  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Titration with HClO4:
    ≥98.5 %
  • GC:
    ≥97.5 %
Frequently Bought Together