Ergocalciferol (vitamin D2, calciferol)
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:50-14-6
:
Formula:C₂₈H₄₄O
MDL Number:MFCD00166988
ADR:6.1,II
:396.66 g/mol
EINECS:200-014-9
UN:2811
Frequently Bought Together
Ethanol absolute, Uvasol® for spectroscopy, Supelco®
Supplier: Merck
Dimethyl sulphoxide ≥99.0%, GPR RECTAPUR®
Supplier: VWR Chemicals
2-Propanol ≥99.8%, HiPerSolv CHROMANORM® for preparative HPLC
Supplier: VWR Chemicals
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Dichloromethane stabilised, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
2,2,4-Trimethylpentane, LiChrosolv® for HPLC, Supelco®
Supplier: Merck
2-Propanol ≥98%, TECHNICAL
Supplier: VWR Chemicals
Methanol, Prepsolv® for preparative chromatography, Supelco®
Supplier: Merck
n-Pentane 98%, Reagent Grade
Supplier: Honeywell Chemicals
Ethyl methanesulphonate 99%
Supplier: Thermo Fisher Scientific
2-Propanol ≥99.8%, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: Avantor
1-Butanol, extra dry 99+%, AcroSeal®
Supplier: Thermo Fisher Scientific
1-Propanol, anhydrous ≥99.9%
Supplier: Thermo Fisher Scientific
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Supplier: Honeywell Chemicals
Petroleum spirit, 100…120 °C, GPR RECTAPUR®
Supplier: VWR Chemicals