Bromate-Bromide concentrate for dilution to 1 L c(Br2) = 0,05 mol/l (0,1 N), Titrisol® volumetric solution, Supelco®
Catalog # 1.09905.0001
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass ampoule
- Pk:1 Ampoul
- Seq:0501
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.11 g/cm3 (20 °C)
- pH:12.8 (H2O, 20 °C)
Specification Test Results
About this item
:
Density:1,11 g/cm3 (20 °C)
:M-Clarity™ quality level = MQ100
:GHS08
:GHS07
Storage Temperature:Ambient
:7758-02-3
Formula:KBrO₃/KBr