Tungstosilicic acid hydrate, EMSURE® for analysis, Supelco®
Catalog # 1.00659.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:100 g
- Seq:3401
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Molar Mass:2878.17 g/mol
- Bulk Density:200 - 500 kg/m3
Specification Test Results
About this item
:
:2896.172 g/mol
:M-Clarity™ quality level = MQ300
EINECS:234-719-8
CAS Number:12027-43-9
Formula:H₄Si(W₃O₁₀)₄.xH₂O
MDL Number:MFCD00036271