


Phenol : chloroform : iso-amyl alcohol (25:24:1) (v/v/v) ≥99%, Biotechnology Grade, low pH
Catalog # 0966-400ML
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:400 mL
- Seq:2665
Specifications
Specification Test Results
- O.D.@ 330 nm:≤0.2
- O.D.@ 405 nm:≤0.1
- O.D.@ 510 nm:≤0.1
- pH @ 25 °C:4.9 – 5.5
- Purity:≥99 %
Specification Test Results
About this item
:
UN:2810
ADR:6.1,II
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
CAS Number:136112-00-0
MDL Number:MFCD00133763