Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
£0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Northern Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Congo Red
Congo red, Sigma-Aldrich®
Congo red, Sigma-Aldrich®
:
SIGMA ALDRICH MICROSCOPY
Congo red, Sigma-Aldrich®
:
SIGMA ALDRICH MICROSCOPY
:
573-58-0
:
:
C₃₂H₂₂N₆Na₂O₆S₂
:
MFCD00004028
:
696.67 g/mol
:
12,02562
:
> 360 °C
:
M-Clarity™ quality level = MQ200
:
209-358-4
:
GHS08
:
Ambient
Merck The Color of Tissue Diagnostics
Frequently Bought Together
VWR® Facial Tissues
: VWR Collection
pH indicator strips, MQuant®, Supelco®
: Merck
VWR® UltraPure, Tris-acetate-EDTA (TAE) buffer
: VWR Chemicals
Hydrochloric acid 1 mol/l (1 N), Titripur® Reag. USP, Reag. Ph. Eur. volumetric solution, Supelco®
: Merck
Test strips, nitrate/nitrite, Dosatest
: VWR Chemicals
pH indicator papers, MQuant®, Supelco®
: Merck
Ammonia solution 25%, EMSURE® ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Test kits, COD cell test, Spectroquant®, Supelco®
: Merck
Acetonitrile ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Test strips for free chlorine, MQuant®, Supelco®
: Merck
Accessories
: Merck
Sodium chloride, EMSURE® ACS, ISO, Reag. Ph. Eur. for analysis, Supelco®
: Merck
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals
Test strips for peroxide, MQuant®, Supelco®
: Merck
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Visual colour kits, total hardness, MQuant®, Supelco®
: Merck
Sodium hydroxide, pellets, EMSURE® for analysis, Supelco®
: Merck