Copper(II) acetate monohydrate 98.0-102.0%, AnalaR NORMAPUR®
84843.230
:
- Pack type:Plastic bottle for solids
- Pk:250 g
- Seq:3545
- Assay:98.0 - 102.0 %
- Insolubility in water (5 %):Max. 0.03 %
- Cl (Chloride):Max. 30 ppm
- SO4 (Sulphate):Max. 100 ppm
- Ca (Calcium):Max. 50 ppm
- Fe (Iron):Max. 20 ppm
- K (Potassium):Max. 100 ppm
- Na (Sodium):Max. 0.05 %
- Ni (Nickel):Max. 100 ppm
:
:13,02651
:115 °C
:1,88 g/cm³ (20 °C)
:9,III
:199.65 g/mol
:205-553-3
:3077
:GHS07
:GHS05
:GHS09
:Ambient
:6046-93-1
:(H₃CCOO)₂Cu·H₂O
:MFCD00149570