2-Methyl-6-(tributylstannyl)pyridine 96%
Catalog # H51567.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:259807-95-9
:
Formula:C₁₈H₃₃NSn
MDL Number:MFCD07787401
:382.16 g/mol
ADR:6.1,III
:GHS06
:GHS08
:GHS07