5-Propyl-1,3-cyclohexanedione
Catalog # APOSOR7071-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
CAS Number:57641-89-1
Formula:C₉H₁₄O₂
MDL Number:MFCD00129456