
Methyl-3-methoxythiophene-2-carboxylate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:62353-75-7
:
Formula:C₇H₈O₃S
MDL Number:MFCD00102115
:172.20 g/mol
Melting Pt:60…63 °C
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together