


2,5-Dichloro-4-(trifluoromethyl)pyridine ≥97%
Catalog # H33442.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:215.99 g/mol
Boiling Pt:76 °C (18 mmHg)
Storage Temperature:Ambient
UN:1993
ADR:3,III
:GHS06
:GHS02
:GHS07
CAS Number:89719-92-6
Formula:C₆H₂Cl₂F₃N
MDL Number:MFCD09749663