Order Entry
Northern Ireland
ContactUsLinkComponent
4-Cyano-4'-hexylbiphenyl ≥97%
Irritant
4-Cyano-4'-hexylbiphenyl ≥97%
Catalog # H55825.06
CAS Number:  
Irritant
4-Cyano-4'-hexylbiphenyl ≥97%
Catalog # H55825.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :263.38 g/mol
Flash Pt:113 °C (235 °F)
EINECS:255-228-5
 :GHS07
Storage Temperature:Ambient
CAS Number:41122-70-7
Formula:C₁₉H₂₁N
MDL Number:MFCD00075144