

4-Chloro-3-(trifluoromethyl)pyridine hydrochloride ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:732306-24-0
:
Formula:C₆H₄Cl₂F₃N
MDL Number:MFCD04972770
:218.01 g/mol
Melting Pt:∼ 135 °C (subl.)
:GHS06
:GHS07
Storage Temperature:Ambient
Frequently Bought Together