3-Butylidenephthalide mixture E/Z 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:551-08-6
:
Formula:C₁₂H₁₂O₂
MDL Number:MFCD00047319
:188.23 g/mol
Boiling Pt:108…109 °C (0,1 mmHg)
Density:1,103
Flash Pt:132 °C (269 °F)
EINECS:208-991-3
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together