
trans-Bis(benzonitrile)dichloropalladium(II) (27,1% Pd)
Catalog # 10006.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:383.57 g/mol
Storage Temperature:Ambient
EINECS:238-085-3
UN:3439
ADR:6.1,III
:GHS07
CAS Number:14220-64-5
Formula:C₁₄H₁₀Cl₂N₂Pd
MDL Number:MFCD00013123