Tris(6,6,7,7,8,8,8-heptafluoro-2,2-dimethyl-3,5-octanedionato-O,O')ytterbium(III) ≥99% (metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18323-96-1
:
Formula:C₃₀H₃₀F₂₁O₆Yb
MDL Number:MFCD00044289
:1058.562 g/mol
Melting Pt:108…111 °C
Storage Temperature:Ambient
EINECS:242-211-2
Related Information
Frequently Bought Together