Tetraethylammonium toluene-4-sulphonate
Catalog # MOLEM88590683
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:301.4500 g/mol
EINECS:211-992-1
CAS Number:733-44-8
Formula:C₁₅H₂₇NO₃S
MDL Number:MFCD00011831