Propargyl-p-toluenesulphonate ≥97%
Catalog # L12075.09
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g
- Seq:3236
Specifications
About this item
:
Boiling Pt:117…121 °C (0,3 mmHg)
Density:1,220
:210.25 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:6165-76-0
:MFCD01462194
Formula:C₁₀H₁₀O₃S
MDL Number:MFCD00078365