
Tetramethylammonium acetate 50% in aqueous solution for synthesis, Sigma-Aldrich®
Catalog # 8.41249.0010
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:10 ml
- Seq:2275
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Density:1.000 g/cm3 (20 °C)
Specification Test Results
About this item
:
:133.20 g/mol
:M-Clarity™ quality level = MQ200
EINECS:234-179-3
:GHS07
Storage Temperature:Ambient
CAS Number:10581-12-1
Formula:C₆H₁₅NO₂
MDL Number:MFCD00011630