Uniform precision stirrers
Supplier: GLASS PRECISION ENGINEERING
Certificates
About this item
These Uni-Form range of all-glass stirrers are made with borosilicate glass precision rod and bearings. The PTFE blades are easily removed or replaced.
- Glass precision and non-whip
- For use with overhead stirrer motors
Frequently Bought Together