ContactUsLinkComponent
Shipping bottles, stainless steel, UN approved, with screw cap, DURAN®
Shipping bottles, stainless steel, UN approved, with screw cap, DURAN®
Catalog # 215-3263
Supplier:  DWK Life Sciences
undefined
Shipping bottles, stainless steel, UN approved, with screw cap, DURAN®
Catalog # 215-3263
Supplier:  DWK Life Sciences
Specifications
  • Thread:
    GL 45
  • Capacity:
    1500 ml
  • Pk:
    1
Specifications
About this item
Manufactured from corrosion resistant AISI Type 316 L (1.4404) stainless steel, and hygienically constructed with all welded seams.
  • Container for shipping hazardous or non hazardous liquids
  • Storage or shipping of precious liquids, such as high purity fine chemicals, pharmaceutical or cosmetic products
  • Storage of light-sensitive liquids
This unbreakable steel bottle is ideal for the storage and safe shipping of dangerous liquid goods such as solvents and reagents without the need for additional protective outer packaging. The bottle has a smooth inner surface finish that corresponds to IIIc (DIN 17441) with rounded inner edges for ease of cleaning. Polished and brushed durable exterior finish. It has a special thread (45 mm) with a built-in pouring lip. Supplied complete with GL 45 stainless steel screw cap and PTFE sealing disc, plus UN certification mark and code.
Certified to UN standards for the carriage of HAZMAT Class 3: Flammable Liquids; classified as dangerous goods in UN packing Group I: (great danger) Groups II (medium danger) and III (low danger). It is suitable for liquids with a relative density of 2,0 or less.
International regulations are subject to change, it is the user’s responsibility to comply with all applicable laws and regulations.
Important note: The supplied DURAN Group stainless steel 45 mm thread closure must be used to maintain UN certification.
With printed Retrace Code (batch identification).