Benzoic acid methyl ester
Catalog # 1.00223.6190
Supplier: Merck
CAS Number:
Specifications
- Pack type:Steel barrel
- Pk:190 L
- Seq:5514
Specifications
About this item
:
:136.15 g/mol
Merck Index:13,06052
Boiling Pt:199,6 °C (1013 hPa)
Melting Pt:–12 °C
Density:1,0888 g/cm³ (20 °C)
Flash Pt:83 °C
Storage Temperature:Ambient
EINECS:202-259-7
:GHS07
CAS Number:93-58-3
Formula:C₆H₅COOCH₃
MDL Number:MFCD00008421