Zinc sulphate concentrate for dilution to 1L, 0.1 M ZnSO4 volumetric solution, Fluka™
Catalog # 32047-1EA
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Ampoule
- Pk:1
- Seq:0001
Specifications
Specification Test Results
- Factor:0.998 - 1.002
Specification Test Results
About this item
:
ADR:9,III
:287.56 g/mol
EINECS:231-793-3
:GHS07
:GHS05
:GHS09
Storage Temperature:Ambient
CAS Number:7446-20-0
Formula:ZnSO₄·7H₂O
MDL Number:MFCD00149894