
3-Fluoro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile 96%
Catalog # H59153.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:247.07 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:624741-47-5
:MFCD18729920
Formula:C₁₃H₁₅BFNO₂