Order Entry
Northern Ireland
ContactUsLinkComponent
Methyl trifluoropyruvate 97%
Corrosive
Flammable
Methyl trifluoropyruvate 97%
Catalog # ACRO379710050
CAS Number:  
undefined
Corrosive
Flammable
Methyl trifluoropyruvate 97%
Catalog # ACRO379710050
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Appearance:
    Clear colorless to light yellow Liquid
  • Infrared spectrum:
    Conforms
  • Refractive index:
    1.3320 to 1.3340 (20°C, 589 nm)
  • GC:
    ≥96.0 %

Specification Test Results

About this item

 :
 :156.06 g/mol
Flash Pt:26 °C
Storage Temperature:Ambient
UN:2924
ADR:3,III
 :GHS05
 :GHS02
CAS Number:13089-11-7
Formula:C₄H₃F₃O₃
MDL Number:MFCD00114936