Order Entry
Northern Ireland
ContactUsLinkComponent
4-(Chloromethyl)benzoic acid
4-(Chloromethyl)benzoic acid
Catalog # MOLEM16061695
Supplier:  Molekula
CAS Number:  
4-(Chloromethyl)benzoic acid
Catalog # MOLEM16061695
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
 :170.60 g/mol
EINECS:216-697-1
CAS Number:1642-81-5
Formula:C₈H₇ClO₂
MDL Number:MFCD00002568