Cerium(III)-2-ethylhexanoate 49% (12% Ce) in 2-ethylhexanoic acid
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Cerium(III) 2-ethylhexanoate
CAS Number:56797-01-4
:
Formula:C₂₄H₄₅CeO₆
MDL Number:MFCD00014004
Density:1.08
Molecular Weight:569.727 g/mol
Flash Pt:>110 °C (230 °F)
EINECS:260-386-3
:GHS08
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together