Order Entry
Northern Ireland
ContactUsLinkComponent
3-Methoxybenzenesulphonamide ≥97%
3-Methoxybenzenesulphonamide ≥97%
Catalog # H28844.03
CAS Number:  
3-Methoxybenzenesulphonamide ≥97%
Catalog # H28844.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

CAS Number:58734-57-9
 :
Formula:C₇H₉NO₃S
MDL Number:MFCD08704579
 :187.22 g/mol
Melting Pt:131…133 °C
 :GHS07
Storage Temperature:Ambient