


Iodomethane 2 M in tert-butyl methyl ether, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:74-88-4
:
Formula:CH₃I
MDL Number:MFCD00001073
:141.94 g/mol
Flash Pt:–18 °C
Storage Temperature:Refrigerator
EINECS:200-819-5
UN:1992
ADR:3,II
:GHS06
:GHS08
:GHS02
Specification Test Results
- Appearance:Clear colorless to light yellow Solution
- Stabilizer:Silver
- Strength:1.90 to 2.20 M
Specification Test Results
Related Information
Frequently Bought Together