Quinoxaline-2,3-dione ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:15804-19-0
:
Formula:C₈H₆N₂O₂
MDL Number:MFCD00006723
:162.15 g/mol
Melting Pt:>300 °C
Storage Temperature:Ambient
EINECS:239-901-0
Related Information
Frequently Bought Together