


Trimethylsilyl isothiocyanate ≥94%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2290-65-5
:
Formula:C₄H₉NSSi
MDL Number:MFCD00004797
:131.27 g/mol
Boiling Pt:143…144 °C
Melting Pt:–33 °C
Density:0,932
Flash Pt:29 °C (84 °F)
EINECS:218-929-7
ADR:3,III
:GHS06
:GHS02
:GHS07
Related Information
Frequently Bought Together