
5-Chloro-1,3-dimethyl-1H-pyrazole ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:54454-10-3
:
Formula:C₅H₇ClN₂
MDL Number:MFCD00051650
:130.58 g/mol
Boiling Pt:153…155 °C
Density:1,137
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together