ContactUsLinkComponent
Pestles, KONTES®, PELLET PESTLES®, KIMBLE®
Catalog # 432-1853
Supplier:  KIMBLEKONTES
undefined
Pestles, KONTES®, PELLET PESTLES®, KIMBLE®
Catalog # 432-1853
Supplier:  KIMBLEKONTES
Specifications
  • Description:
    Pistils, polypropylene, 100 pcs
  • Length:
    70 mm
  • Pk:
    100
Specifications
About this item
Grinders are ideal for resuspending protein and DNA pellets or grinding soft tissues. Pestles are specially designed to fit either 0,5 or 1,5 ml microtubes.
Disposable, autoclavable polypropylene pestles eliminate the need for pretreatment of pestles and microtubes. The CTFE/stainless steel pestles are reusable.
Ordering information: Pestle may be hand-operated, or driven by optional cordless mixer motor. Mixer motor includes two AA size batteries and adapter for attaching pestle.