Tubing, LDPE, fine-bore
Supplier: Smiths Medical
Certificates
About this item
PE, translucent.
- Lightweight and flexible
- High chemical resistance
- Smooth bore, easily cleaned
This tubing has surgical non-toxic quality, minimises trauma when used in vivo. Suitable for sheathing delicate glass fibres in telecommunications.
Frequently Bought Together