Hydroxylamine solution, alkaline R1 (A+B) Reag. Ph. Eur. 1044303
Catalog # 87849.220
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:200 mL
- Seq:2530
Specifications
About this item
:
:33.03 g/mol
EINECS:0
CAS Number:7803-49-8
Formula:H₃NO
MDL Number:MFCD00044522