Order Entry
Northern Ireland
ContactUsLinkComponent
Hydroxylamine solution, alcoholic Reag. Ph. Eur. 1044301
Hydroxylamine solution, alcoholic Reag. Ph. Eur. 1044301
Catalog # 87847.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Hydroxylamine solution, alcoholic Reag. Ph. Eur. 1044301
Catalog # 87847.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :33.03 g/mol
CAS Number:7803-49-8
Formula:H₃NO
MDL Number:MFCD00044522