Order Entry
Northern Ireland
ContactUsLinkComponent
Dithizone solution R2 Reag. Ph. Eur. 1033903
Dithizone solution R2 Reag. Ph. Eur. 1033903
Catalog # 87827.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Dithizone solution R2 Reag. Ph. Eur. 1033903
Catalog # 87827.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :256.33 g/mol
CAS Number:60-10-6
Formula:C₆H₅NHNHCSN=NC₆H₅
MDL Number:MFCD00003025