Order Entry
Northern Ireland
ContactUsLinkComponent
3-(Methylsulphonyl)benzoic acid 95%
3-(Methylsulphonyl)benzoic acid 95%
Catalog # ACRO424860025
CAS Number:  
undefined
3-(Methylsulphonyl)benzoic acid 95%
Catalog # ACRO424860025
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    2,5 g
  • Seq:
    3122

Specifications

Specification Test Results

  • Appearance:
    White to light yellow Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    230 to 238 °C
  • HPLC:
    ≥94 %

Specification Test Results

About this item

 :
 :200.21 g/mol
Melting Pt:230…238 °C
Storage Temperature:Ambient
CAS Number:5345-27-7
Formula:C₈H₈O₄S
MDL Number:MFCD00216488