4,4-Dimethyl-3-thiosemicarbazide ≥95%
Catalog # L10337.06
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
CAS Number:6926-58-5
:
Formula:C₃H₉N₃S
MDL Number:MFCD00041308
:119.1887 g/mol
Melting Pt:142…147 °C
ADR:,