Order Entry
Northern Ireland
ContactUsLinkComponent
4-(3-Chlorophenyl)-3-thiosemicarbazide ≥98%
Toxic
4-(3-Chlorophenyl)-3-thiosemicarbazide ≥98%
Catalog # L11610.06
CAS Number:  
Toxic
4-(3-Chlorophenyl)-3-thiosemicarbazide ≥98%
Catalog # L11610.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

CAS Number:42135-76-2
 :
Formula:C₇H₈ClN₃S
MDL Number:MFCD00041287
 :201.68 g/mol
Melting Pt:130…133 °C
Storage Temperature:Ambient
ADR:6.1,III
 :GHS06