Order Entry
Northern Ireland
ContactUsLinkComponent
4'-Ethoxyacetanilide, Supelco®
4'-Ethoxyacetanilide, Supelco®
Catalog # PHR1094-1G
Supplier:  Merck
CAS Number:  
undefined
4'-Ethoxyacetanilide, Supelco®
Catalog # PHR1094-1G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :179.22 g/mol
EINECS:200-533-0
CAS Number:62-44-2
Formula:C₁₀H₁₃NO₂
MDL Number:MFCD00009094