


Phenol liquified ≥90% for synthesis
Supplier: Merck
Certificates
About this item
CAS Number:108-95-2
:
Formula:C₆H₅OH
MDL Number:MFCD00002143
:94.11 g/mol
ADR:6.1,II
:M-Clarity™ quality level = MQ200
EINECS:203-632-7
UN:2821
:GHS06
:GHS05
:GHS08
Storage Temperature:Ambient
Specification Test Results
- Density:1.07 g/cm³ (25 °C)
- Flash point:81 °C Information refers to the main component.
- pH value:(H₂O) acidic
- WGK:WGK 2 water endangering
- R Phrase:R 23/24/25-34-68-48/20/21/22
- S Phrase:S 24/25-26-28-36/37/39-45
Specification Test Results
Frequently Bought Together