
3,5-Dichloro-2-hydroxybenzenesulphonic acid sodium salt for synthesis, Sigma-Aldrich®
: Merck
:54970-72-8
:
:C₆H₃Cl₂NaO₄S
:MFCD00009798
:265.05 g/mol
:M-Clarity™ quality level = MQ200
:259-416-8
:GHS07
:Ambient
- Solubility:(20 °C) soluble
- Melting Point:300 °C
- Molar Mass:265.05 g/mol
- pH:7 (53 g/l, H2O, 20 °C)
Frequently Bought Together