
Diethyl aminomalonate hydrochloride 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13433-00-6
:
Formula:C₇H₁₄ClNO₄
MDL Number:MFCD00012510
:211.65 g/mol
Melting Pt:∼ 165 °C (dec.)
EINECS:236-556-8
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together