
[2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphonium bromide ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:69891-92-5
:
Formula:C₂₄H₂₆BrO₂P
MDL Number:MFCD00012000
:457.36 g/mol
Melting Pt:183…195 °C
EINECS:274-189-5
:GHS07
Storage Temperature:Ambient
Related Information
Frequently Bought Together