ContactUsLinkComponent
Glass density bottles acc. Gay-Lussac
Glass density bottles acc. Gay-Lussac
Catalog # WITG3.902.002
Supplier:  witeg Labortechnik
undefined
Glass density bottles acc. Gay-Lussac
Catalog # WITG3.902.002
Supplier:  witeg Labortechnik
Specifications
  • Volume (nom.):
    2 ±0,006 ml
  • Environmentally Preferable:
  • Pk:
    1
Specifications
About this item
Made of borosilicate glass.
Pycnometer with with ground-in capillary stopper, precisely adjusted to the second place of decimal, acc. to ISO 3507.