



Heptane (mixture of isomers), BAKER, Laboratory Reagent, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:142-82-5
:
Formula:H₃C(CH₂)₅CH₃
MDL Number:MFCD00009544
Merck Index:13,04679
Boiling Pt:97…98 °C (1013 hPa)
Melting Pt:–90,5 °C
Density:0,68 g/cm³ (20 °C)
Flash Pt:–4 °C
UN:1206
ADR:3,II
:100.2 g/mol
EINECS:205-563-8
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
Specification Test Results
- Boiling Range:94 - 99 °C
Specification Test Results
Frequently Bought Together