Order Entry
Northern Ireland
ContactUsLinkComponent
4'-Hydroxyacetophenone oxime
4'-Hydroxyacetophenone oxime
Catalog # PROOMM0042.12
CAS Number:  
4'-Hydroxyacetophenone oxime
Catalog # PROOMM0042.12
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
CAS Number:34523-34-7
Formula:C₈H₉NO₂
MDL Number:MFCD00481697