Order Entry
Northern Ireland
ContactUsLinkComponent
3'-Phenoxyacetophenone
3'-Phenoxyacetophenone
Catalog # APOSOR8954-1G
Supplier:  Apollo Scientific
CAS Number:  
3'-Phenoxyacetophenone
Catalog # APOSOR8954-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :212.2439 g/mol
EINECS:251-259-3
CAS Number:32852-92-9
Formula:C₁₄H₁₂O₂
MDL Number:MFCD08543460